Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Cntnap5b
- Entrez Gene ID
-
241175
- Gene Name
- contactin associated protein-like 5B
- Gene Aliases
-
C230078M14Rik, Caspr5-2
- Chromosome Location
-
Chr.1: 99772247 - 100486066 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_172851.2 |
NP_766439.2 |
3 - 4 |
383 |
77 |
View Full Product Details