Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Lgr6
Entrez Gene ID
329252
Gene Name
leucine-rich repeat-containing G protein-coupled receptor 6
Gene Aliases
A530037C04Rik, D830026M09
Chromosome Location
Chr.1: 134983300 - 135105374 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033409.3 NP_001028581.1 13 - 14 1347 53
GenBank mRNA AK052873.1 - 8 - 9 547 53
AK085901.1 - 13 - 14 1306 53
AK137380.1 - 13 - 14 1348 53
BC141210.1 - 13 - 14 1299 53

View Full Product Details