Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gpr82
Entrez Gene ID
319200
Gene Name
G protein-coupled receptor 82
Gene Aliases
-
Chromosome Location
Chr.X: 13658153 - 13667433 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_175669.3 NP_783600.1 1 - 2 75 124
GenBank mRNA AK033761.1 - 1 - 2 77 124
BC116704.1 - 1 - 2 70 124
FJ789815.1 - 1 - 2 112 124

View Full Product Details