The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013529.3 | NP_038557.1 | 1 - 2 | 94 | 126 |
GenBank mRNA | AK053670.1 | - | 1 - 2 | 72 | 126 |
BC031928.1 | - | 1 - 2 | 62 | 126 | |
CJ163487.1 | - | 1 - 2 | 96 | 126 |