Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Zbtb37
Entrez Gene ID
240869
Gene Name
zinc finger and BTB domain containing 37
Gene Aliases
D430004I08Rik
Chromosome Location
Chr.1: 161008792 - 161034864 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK078748.1 - 1 - 2 60 57

View Full Product Details