The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001033039.2 | NP_001028211.1 | 1 - 2 | 665 | 88 |
GenBank mRNA | AK076812.1 | - | 1 - 2 | 388 | 88 |
AK140290.1 | - | 1 - 2 | 666 | 88 | |
BC141196.1 | - | 1 - 2 | 578 | 88 |