Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Chil5
Entrez Gene ID
229687
Gene Name
chitinase-like 5
Gene Aliases
Bclp, Bclp1, Bclp2, Chi3l7
Chromosome Location
Chr.3: 106016919 - 106033131 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC116403.1 - 5 - 6 616 121

View Full Product Details