Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dnm3
Entrez Gene ID
103967
Gene Name
dynamin 3
Gene Aliases
9630020E24Rik, AW061159, B230343F03Rik, C530045C17, mKIAA0820
Chromosome Location
Chr.1: 161982450 - 162478323 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001038619.1 NP_001033708.1 2 - 3 440 117
NM_172646.2 NP_766234.1 2 - 3 444 117
GenBank mRNA AK035951.1 - 2 - 3 423 117
AK036199.1 - 2 - 3 405 117
AK049724.1 - 2 - 3 445 117
AK082698.1 - 2 - 3 384 117
AK147678.1 - 2 - 3 384 117
BC141143.1 - 2 - 3 288 117
BC141144.1 - 2 - 3 288 117

View Full Product Details