Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tekt4
Entrez Gene ID
71840
Gene Name
tektin 4
Gene Aliases
1700010L19Rik
Chromosome Location
Chr.17: 25433645 - 25476602 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_027951.1 NP_082227.1 2 - 3 740 96
GenBank mRNA AK005842.1 - 2 - 3 742 96
AY485267.1 - 2 - 3 740 96
BC117510.1 - 2 - 3 693 96
BC117526.1 - 2 - 3 693 96

View Full Product Details