The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001004359.2 | NP_001004359.1 | 2 - 3 | 364 | 81 |
NM_001005385.1 | NP_001005385.1 | 3 - 4 | 445 | 81 | |
NM_026081.5 | NP_080357.4 | 2 - 3 | 364 | 81 | |
GenBank mRNA | AK008944.1 | - | 3 - 4 | 358 | 81 |
AK083192.1 | - | 3 - 4 | 405 | 81 | |
AK170837.1 | - | 2 - 3 | 322 | 81 | |
BC070413.1 | - | 2 - 3 | 312 | 81 | |
BC085157.1 | - | 3 - 4 | 347 | 81 |