Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Cdk20
- Entrez Gene ID
-
105278
- Gene Name
- cyclin-dependent kinase 20
- Gene Aliases
-
4932702G04Rik, AU019450, AW558027, CDCH, Ccrk, PNQLARE, p42
- Chromosome Location
-
Chr.13: 64432363 - 64439722 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AY005133.1 |
- |
3 - 4 |
429 |
63 |
View Full Product Details