Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Dhrs3
- Entrez Gene ID
-
20148
- Gene Name
- dehydrogenase/reductase (SDR family) member 3
- Gene Aliases
-
Rsdr1, retSDR1
- Chromosome Location
-
Chr.4: 144892609 - 144927645 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC008980.1 |
- |
3 - 4 |
354 |
67 |
View Full Product Details