The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001013759.2 | NP_001013781.1 | 3 - 4 | 829 | 109 |
GenBank mRNA | BC147480.1 | - | 3 - 4 | 829 | 109 |
BC150847.1 | - | 3 - 4 | 829 | 109 |