Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Wsb1
Entrez Gene ID
78889
Gene Name
WD repeat and SOCS box-containing 1
Gene Aliases
1110056B13Rik, 2700038M07Rik
Chromosome Location
Chr.11: 79239382 - 79254700 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK168468.1 - 5 - 6 992 80
AK168565.1 - 5 - 6 992 80

View Full Product Details