Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Map3k1
Entrez Gene ID
26401
Gene Name
mitogen-activated protein kinase kinase kinase 1
Gene Aliases
MAPKKK1, MEKK1, Mekk
Chromosome Location
Chr.13: 111746433 - 111818269 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011945.2 NP_036075.2 13 - 14 2376 84
GenBank mRNA AF117340.1 - 13 - 14 2376 84
L13103.1 - 3 - 4 385 84

View Full Product Details