Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Cdc14b
- Entrez Gene ID
-
218294
- Gene Name
- CDC14 cell division cycle 14B
- Gene Aliases
-
2810432N10Rik, A530086E13Rik, AA472821, CDC14B3, Cdc14B1
- Chromosome Location
-
Chr.13: 64189271 - 64275290 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK183075.1 |
- |
2 - 3 |
147 |
87 |
View Full Product Details