Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Msl1
- Entrez Gene ID
-
74026
- Gene Name
- male-specific lethal 1 homolog (Drosophila)
- Gene Aliases
-
2810017F12Rik, 4121402D02Rik, 4930463F05Rik, AA682082, Msl-1
- Chromosome Location
-
Chr.11: 98795468 - 98807862 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK015496.1 |
- |
3 - 4 |
699 |
71 |
AK053719.1 |
- |
3 - 4 |
1405 |
71 |
View Full Product Details