The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001045527.1 | NP_001038992.1 | 5 - 6 | 1912 | 89 |
GenBank mRNA | AK136195.1 | - | 2 - 3 | 378 | 89 |
AK166561.1 | - | 2 - 3 | 354 | 89 |