The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001170954.1 | NP_001164425.1 | 1 - 2 | 117 | 56 |
GenBank mRNA | AK162529.1 | - | 1 - 2 | 116 | 56 |
BY770781.1 | - | 1 - 2 | 119 | 56 |