The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001205343.1 | NP_001192272.1 | 26 - 27 | 3651 | 62 |
NM_130887.3 | NP_570957.2 | 25 - 26 | 3585 | 62 | |
GenBank mRNA | AF314171.1 | - | 25 - 26 | 3558 | 62 |
AK053198.1 | - | 26 - 27 | 3653 | 62 | |
AK086767.1 | - | 25 - 26 | 3568 | 62 | |
BC005747.1 | - | 7 - 8 | 1122 | 62 | |
BC132475.1 | - | 25 - 26 | 3525 | 62 | |
BC137854.1 | - | 25 - 26 | 3525 | 62 | |
BC144975.1 | - | 26 - 27 | 3591 | 62 |