The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_010465.2 | NP_034595.2 | 1 - 2 | 446 | 53 |
GenBank mRNA | AB221685.1 | - | 1 - 2 | 402 | 53 |
AK050798.1 | - | 1 - 2 | 214 | 53 | |
BC028482.1 | - | 1 - 2 | 446 | 53 | |
BC141061.1 | - | 1 - 2 | 425 | 53 | |
BX528590.1 | - | 1 - 2 | 447 | 53 | |
X12504.1 | - | 2 - 3 | 471 | 53 | |
X16510.1 | - | - | 407 | 53 | |
X16511.1 | - | 1 - 2 | 529 | 53 |