Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tubgcp6
Entrez Gene ID
328580
Gene Name
tubulin, gamma complex associated protein 6
Gene Aliases
AI843275, D430023H11, mKIAA1669
Chromosome Location
Chr.15: 89099097 - 89123208 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001163319.1 NP_001156791.1 14 - 15 2731 71
GenBank mRNA CK637726.1 - 3 - 4 317 71

View Full Product Details