Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Kctd4
Entrez Gene ID
67516
Gene Name
potassium channel tetramerisation domain containing 4
Gene Aliases
2210017A09Rik, AU017169
Chromosome Location
Chr.14: 75955003 - 75965217 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK163645.1 - - 764 86

View Full Product Details