Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cnst
Entrez Gene ID
226744
Gene Name
consortin, connexin sorting protein
Gene Aliases
9630058J23Rik
Chromosome Location
Chr.1: 179546349 - 179627473 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_146105.3 NP_666217.2 2 - 3 522 128
GenBank mRNA AK036332.1 - 2 - 3 554 128
AK087361.1 - 2 - 3 540 128
AK146681.1 - 2 - 3 612 128
BC054802.1 - 2 - 3 522 128

View Full Product Details