The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_173393.2 | NP_775569.2 | 2 - 3 | 953 | 65 |
GenBank mRNA | AY534252.1 | - | 2 - 3 | 659 | 65 |
BC024502.1 | - | 2 - 3 | 509 | 65 |