Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Mettl3
Entrez Gene ID
56335
Gene Name
methyltransferase like 3
Gene Aliases
2310024F18Rik, M6A, Spo8
Chromosome Location
Chr.14: 52294841 - 52305124 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019721.2 NP_062695.2 2 - 3 455 78
GenBank mRNA AK035061.1 - 2 - 3 449 78
AK085189.1 - 2 - 3 442 78
AK089158.1 - 2 - 3 78
AK151616.1 - 2 - 3 442 78
AK153113.1 - 2 - 3 442 78
BC012526.1 - 2 - 3 439 78

View Full Product Details