The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001014996.2 | NP_001014996.2 | 4 - 5 | 998 | 95 |
GenBank mRNA | AK077045.1 | - | 4 - 5 | 998 | 95 |
BC092399.1 | - | 4 - 5 | 984 | 95 |