The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_201639.2 | NP_964001.2 | 3 - 4 | 1124 | 70 |
NM_207663.3 | NP_997546.2 | 3 - 4 | 1124 | 70 | |
GenBank mRNA | AJ579700.1 | - | 3 - 4 | 1075 | 70 |
AJ579701.1 | - | 3 - 4 | 1075 | 70 | |
AK053514.1 | - | 2 - 3 | 142 | 70 | |
AK143019.1 | - | 3 - 4 | 1125 | 70 | |
AK172937.1 | - | 2 - 3 | 205 | 70 | |
BC026872.1 | - | 4 - 5 | 968 | 70 | |
BC040093.1 | - | 4 - 5 | 968 | 70 |