Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rnpc3
Entrez Gene ID
67225
Gene Name
RNA-binding region (RNP1, RRM) containing 3
Gene Aliases
2810441O16Rik, AI447568, C030014B17Rik
Chromosome Location
Chr.3: 113605067 - 113630149 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001038696.1 NP_001033785.1 6 - 7 713 103
NM_001287015.1 NP_001273944.1 6 - 7 713 103
NM_026043.3 NP_080319.2 6 - 7 713 103
GenBank mRNA AK013288.1 - 6 - 7 657 103
AK134241.1 - 6 - 7 678 103
AK161063.1 - 6 - 7 696 103
AK161974.1 - 6 - 7 666 103
AK163729.1 - 6 - 7 666 103
AK173272.1 - 6 - 7 464 103
BC016603.1 - 3 - 4 229 103
BC043695.1 - 6 - 7 682 103

View Full Product Details