The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_010881.2 | NP_035011.1 | 21 - 22 | 4887 | 98 |
GenBank mRNA | BC068177.1 | - | 18 - 19 | 4195 | 98 |
BC080866.1 | - | 21 - 22 | 4564 | 98 | |
U56920.1 | - | 20 - 21 | 4445 | 98 | |
U64828.1 | - | 21 - 22 | 4800 | 98 |