The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001164107.1 | NP_001157579.1 | 2 - 3 | 371 | 94 |
NM_001164108.1 | NP_001157580.1 | 2 - 3 | 469 | 94 | |
NM_019955.2 | NP_064339.2 | 2 - 3 | 388 | 94 | |
GenBank mRNA | AF178953.1 | - | 2 - 3 | 317 | 94 |
AK012177.1 | - | 2 - 3 | 371 | 94 | |
AK154505.1 | - | 2 - 3 | 381 | 94 | |
BC029210.1 | - | 2 - 3 | 339 | 94 |