The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001164107.1 | NP_001157579.1 | 4 - 5 | 834 | 69 |
NM_001164108.1 | NP_001157580.1 | 4 - 5 | 932 | 69 | |
NM_019955.2 | NP_064339.2 | 4 - 5 | 851 | 69 |