The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_177049.5 | NP_796023.2 | 1 - 2 | 941 | 78 |
GenBank mRNA | AB099712.1 | - | 1 - 2 | 284 | 78 |
AK033956.1 | - | 1 - 2 | 574 | 78 | |
AK034118.1 | - | 1 - 2 | 574 | 78 | |
AK036161.1 | - | 1 - 2 | 587 | 78 | |
AY341261.1 | - | 1 - 2 | 941 | 78 |