The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019955.2 | NP_064339.2 | 1 - 2 | 245 | 67 |
GenBank mRNA | AF178953.1 | - | 1 - 2 | 174 | 67 |
AK154505.1 | - | 1 - 2 | 238 | 67 | |
BC029210.1 | - | 1 - 2 | 196 | 67 | |
BY209344.1 | - | 1 - 2 | 206 | 67 |