Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Kctd3
Entrez Gene ID
226823
Gene Name
potassium channel tetramerisation domain containing 3
Gene Aliases
4930438A20Rik, 9330185B06, E330032J19Rik, NY-REN-45
Chromosome Location
Chr.1: 188971095 - 189007868 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172650.2 NP_766238.1 8 - 9 856 56
GenBank mRNA AK033491.1 - 8 - 9 857 56
AK034380.1 - 8 - 9 856 56
AK049362.1 - 8 - 9 854 56
AK139700.1 - 8 - 9 856 56
BC061458.1 - 8 - 9 844 56
BC079581.1 - 8 - 9 843 56

View Full Product Details