Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tc2n
Entrez Gene ID
74413
Gene Name
tandem C2 domains, nuclear
Gene Aliases
4933406D09Rik, Mtac2d1, Tac2-N
Chromosome Location
Chr.12: 101645446 - 101719000 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001082976.2 NP_001076445.1 7 - 8 901 154
NM_001286364.1 NP_001273293.1 7 - 8 1564 154
NM_028924.3 NP_083200.1 7 - 8 1253 154
GenBank mRNA AB062282.1 - 6 - 7 733 154
AK016683.1 - 7 - 8 1585 154
AK136558.1 - 7 - 8 902 154
BC120793.1 - 7 - 8 1253 154

View Full Product Details