Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Aloxe3
Entrez Gene ID
23801
Gene Name
arachidonate lipoxygenase 3
Gene Aliases
e-LOX-3, eLOX-3
Chromosome Location
Chr.11: 69125674 - 69149115 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011786.2 NP_035916.2 11 - 12 1586 76

View Full Product Details