The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001029936.3 | NP_001025107.1 | 5 - 6 | 2252 | 58 |
NM_001281818.1 | NP_001268747.1 | 7 - 8 | 2469 | 58 | |
GenBank mRNA | AK220465.1 | - | 5 - 6 | 2179 | 58 |