The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007760.3 | NP_031786.2 | 13 - 14 | 2073 | 68 |
GenBank mRNA | AK041431.1 | - | 13 - 14 | 1848 | 68 |
AK132179.1 | - | 14 - 15 | 1900 | 68 | |
AK170740.1 | - | 13 - 14 | 1859 | 68 | |
BC006668.1 | - | 13 - 14 | 1820 | 68 | |
X85983.1 | - | 13 - 14 | 1791 | 68 |