The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_133922.3 | NP_598683.3 | 5 - 6 | 864 | 94 |
GenBank mRNA | AK040525.1 | - | 5 - 6 | 866 | 94 |
AK128931.1 | - | 5 - 6 | 815 | 94 | |
AK162934.1 | - | 5 - 6 | 572 | 94 | |
AK173281.1 | - | 5 - 6 | 2568 | 94 | |
BC066816.1 | - | 5 - 6 | 784 | 94 |