Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Yme1l1
Entrez Gene ID
27377
Gene Name
YME1-like 1 (S. cerevisiae)
Gene Aliases
FtsH1, Ftsh
Chromosome Location
Chr.2: 23156505 - 23199260 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013771.5 NP_038799.1 13 - 14 1606 98
GenBank mRNA AF090430.1 - 13 - 14 1584 98
AK045141.1 - 13 - 14 1591 98
AK079175.1 - 13 - 14 1566 98
AK137665.1 - 13 - 14 1554 98
AY136286.1 - 13 - 14 1422 98
AY136287.1 - 13 - 14 1575 98
BC007128.1 - 13 - 14 1567 98

View Full Product Details