Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rpl12
Entrez Gene ID
269261
Gene Name
ribosomal protein L12
Gene Aliases
E430018F03
Chromosome Location
Chr.2: 32961712 - 32964045 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009076.3 NP_033102.2 4 - 5 384 76
GenBank mRNA AK002973.1 - 4 - 5 383 76
AK008347.1 - 4 - 5 383 76
AK012349.1 - 4 - 5 383 76
AK012428.1 - 4 - 5 381 76
AK088464.1 - 4 - 5 383 76
AK144905.1 - 4 - 5 383 76
AK150318.1 - 4 - 5 384 76
AK151532.1 - 4 - 5 383 76
AK151592.1 - 4 - 5 383 76
AK167761.1 - 4 - 5 403 76
AK168121.1 - 4 - 5 383 76
AK198056.1 - 2 - 3 144 76
BC018321.1 - 4 - 5 380 76
BC081469.1 - 4 - 5 367 76
BC090393.1 - - 362 76
L04280.1 - 4 - 5 371 76

View Full Product Details