Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Acvr2a
Entrez Gene ID
11480
Gene Name
activin receptor IIA
Gene Aliases
ActrIIa, Acvr2, TactrII
Chromosome Location
Chr.2: 48814109 - 48903264 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_007396.4 NP_031422.3 4 - 5 1198 78
GenBank mRNA AK041246.1 - 4 - 5 1151 78
BC138823.1 - 4 - 5 859 78
M65287.1 - 4 - 5 605 78

View Full Product Details