Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gtpbp4
Entrez Gene ID
69237
Gene Name
GTP binding protein 4
Gene Aliases
2610028C09Rik, Crfg, Gtpbp3, NGB, Nog1
Chromosome Location
Chr.13: 8972464 - 8996012 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_027000.4 NP_081276.2 4 - 5 483 95
GenBank mRNA AF325354.1 - 4 - 5 467 95
AF348208.1 - 4 - 5 490 95
AK011580.1 - 4 - 5 494 95
AK141852.1 - 4 - 5 493 95
AK163372.1 - 3 - 4 290 95
AK164330.1 - 4 - 5 484 95
AK166711.1 - 4 - 5 489 95
BC005514.1 - 4 - 5 464 95
BC025431.1 - 4 - 5 488 95

View Full Product Details