Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gimap5
Entrez Gene ID
317757
Gene Name
GTPase, IMAP family member 5
Gene Aliases
D630024P16, E230026N22Rik
Chromosome Location
Chr.6: 48746137 - 48755235 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_175035.5 NP_778200.1 1 - 2 232 78
GenBank mRNA AB126961.1 - 1 - 2 232 78
AK052694.1 - 1 - 2 165 78
BC095995.1 - 1 - 2 144 78
BU694320.1 - 1 - 2 190 78

View Full Product Details