The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | RNADB ID | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|---|
RefSeq | NR_015348.1 | 4 - 5 | 1156 | 72 | ||
GenBank mRNA | AK133929.1 | - | 2 - 3 | 377 | 72 | |
U20366.1 | - | LIT1565 Hoxa11as |
4 - 5 | 1156 | 72 | |
U20367.1 | - | LIT1564 Hoxa11as |
2 - 3 | 724 | 72 | |
U20369.1 | - | LIT1563 Hoxa11as |
2 - 3 | 361 | 72 |
Gene Name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromosome | |
---|---|---|---|---|---|---|
Upstream | homeobox A11 | Hoxa11 | Hox-1.9, Hoxa-11 | -853.0 | Anti-sense | Chr. - 52242105 - 52245810 on Build GRCm38 |
Downstream | homeobox A13 | Hoxa13 | Hd, Hox-1.10 | 9.4kb | Anti-sense | Chr. - 52258853 - 52260880 on Build GRCm38 |