Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pygb
Entrez Gene ID
110078
Gene Name
brain glycogen phosphorylase
Gene Aliases
-
Chromosome Location
Chr.2: 150786796 - 150831748 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_153781.1 NP_722476.1 6 - 7 850 62
GenBank mRNA AK032769.1 - 6 - 7 863 62
AK035673.1 - 6 - 7 580 62
AK134667.1 - 6 - 7 860 62
AK142716.1 - 6 - 7 860 62
AK147763.1 - 6 - 7 882 62
AK168381.1 - 6 - 7 861 62
AK169052.1 - 6 - 7 885 62
AK169631.1 - 6 - 7 864 62
BC031457.1 - 6 - 7 858 62
BC035283.1 - 6 - 7 850 62

View Full Product Details