Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cage1
Entrez Gene ID
71213
Gene Name
cancer antigen 1
Gene Aliases
4933427I01Rik, Ctag3
Chromosome Location
Chr.13: 37994314 - 38036937 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_027724.2 NP_082000.2 9 - 10 2497 81
GenBank mRNA AK132733.1 - 5 - 6 1046 81
AY695827.1 - 8 - 9 2232 81

View Full Product Details