The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001110224.1 | NP_001103694.1 | 6 - 7 | 1214 | 57 |
NM_010025.2 | NP_034155.2 | 6 - 7 | 1199 | 57 | |
GenBank mRNA | AK014264.1 | - | 6 - 7 | 1200 | 57 |
AK164119.1 | - | 6 - 7 | 1101 | 57 | |
BC056391.1 | - | 6 - 7 | 1107 | 57 | |
BC057010.1 | - | 6 - 7 | 1199 | 57 | |
BC062974.1 | - | 6 - 7 | 1199 | 57 |