Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Celf1
Entrez Gene ID
13046
Gene Name
CUGBP, Elav-like family member 1
Gene Aliases
1600010O03Rik, AA407467, Brunol2, CUG-BP, CUG-BP1, CUGBP, Cugbp1, D2Wsu101e, HNAB50, NAB50
Chromosome Location
Chr.2: 90940387 - 91019497 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001244891.1 NP_001231820.1 3 - 4 488 71
NM_001244903.1 NP_001231832.1 2 - 3 435 71
NM_017368.3 NP_059064.2 3 - 4 587 71
NM_198683.2 NP_941955.1 2 - 3 435 71
GenBank mRNA AF267535.1 - 1 - 2 191 71
AF314172.1 - 1 - 2 29 71
AJ007987.1 - 1 - 2 180 71
AK014492.1 - 2 - 3 348 71
AK143698.1 - 3 - 4 389 71
AK156725.1 - 2 - 3 348 71
AK161213.1 - 2 - 3 395 71
BC021393.1 - 2 - 3 300 71
BU056861.1 - 2 - 3 435 71
BU923429.1 - 2 - 3 290 71
CJ092744.1 - 3 - 4 530 71

View Full Product Details